Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SMT1
Entrez Gene ID
831216
Gene Name
sterol methyltransferase 1
Gene Aliases
AT5G13710, CEPHALOPOD, CPH, MSH12.18, MSH12_18, SAM:CYCLOARTENOL-C24-METHYLTRANSFERASE, sterol methyltransferase 1
Chromosome Location
Chr.5: 4423555 - 4427324 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001085110.1 NP_001078579.1 5 - 6 435 65
NM_121374.4 NP_196875.1 6 - 7 719 65
GenBank mRNA AF090372.1 - 6 - 7 386 65
AF195648.1 - 6 - 7 397 65
AY120716.1 - 6 - 7 411 65
BT000058.1 - 5 - 6 344 65

View Full Product Details