Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ARFA1B
Entrez Gene ID
831319
Gene Name
ADP-ribosylation factor A1B
Gene Aliases
ADP-ribosylation factor A1B, AT5G14670, ATARFA1B, T15N1.160, T15N1_160
Chromosome Location
Chr.5: 4728947 - 4730952 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001343353.1 NP_001318560.1 2 - 3 200 86
NM_001343354.1 NP_001331344.1 2 - 3 209 86
NM_001343355.1 NP_001331345.1 2 - 3 276 86

View Full Product Details