Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AO
Entrez Gene ID
831328
Gene Name
L-aspartate oxidase
Gene Aliases
AT5G14760, FIN4, FLAGELLIN-INSENSITIVE 4, L-aspartate oxidase, T9L3.60, T9L3_60
Chromosome Location
Chr.5: 4768263 - 4772153 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_121480.4 NP_568304.1 5 - 6 1282 93
GenBank mRNA AY045626.1 - 5 - 6 941 93
BT001009.1 - 4 - 5 785 93

View Full Product Details