Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KAN
Entrez Gene ID
831518
Gene Name
Homeodomain-like superfamily protein
Gene Aliases
AT5G16560, KAN1, KANADI, KANADI 1, MQK4.31, MQK4_31
Chromosome Location
Chr.5: 5406863 - 5411766 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_121662.3 NP_568334.1 1 - 2 1406 68
GenBank mRNA AY030192.1 - 1 - 2 1064 68
AY048688.1 - 1 - 2 741 68
BT026436.1 - 1 - 2 732 68

View Full Product Details