Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IRX3
Entrez Gene ID
831608
Gene Name
Cellulose synthase family protein
Gene Aliases
AT5G17420, ATCESA7, CELLULOSE SYNTHASE CATALYTIC SUBUNIT 7, CELLULOSE SYNTHASE CATALYTIC SUBUNIT, CELLULOSE SYNTHASE CATALYTIC SUBUNIT 7, CESA7, IRREGULAR XYLEM 3, MUR10, MURUS 10, T10B6.80, T10B6_80
Chromosome Location
Chr.5: 5736630 - 5741500 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_121748.4 NP_197244.1 4 - 5 738 73
GenBank mRNA AY139754.1 - 4 - 5 690 73
BT004543.1 - 4 - 5 645 73
KJ138681.1 - 4 - 5 645 73

View Full Product Details