Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- MPK16
- Entrez Gene ID
-
832019
- Gene Name
- mitogen-activated protein kinase 16
- Gene Aliases
-
AT5G19010, T16G12.50, T16G12_50, mitogen-activated protein kinase 16
- Chromosome Location
-
Chr.5: 6344770 - 6348267 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_121906.4 |
NP_197402.1 |
5 - 6 |
1358 |
81 |
View Full Product Details