Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ECR1
Entrez Gene ID
832038
Gene Name
E1 C-terminal related 1
Gene Aliases
AT5G19180, E1 C-terminal related 1, T24G5.80, T24G5_80
Chromosome Location
Chr.5: 6453248 - 6456033 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_121923.4 NP_568370.1 8 - 9 1206 69
GenBank mRNA AK226388.1 - 8 - 9 1180 69
AY050426.1 - 8 - 9 1143 69
AY120692.1 - 8 - 9 1079 69

View Full Product Details