Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SYCO ARATH
Entrez Gene ID
817673
Gene Name
Cysteinyl-tRNA synthetase, class Ia family protein
Gene Aliases
AT2G31170, FIONA, T16B12.16, cysteinyl t-RNA synthetase
Chromosome Location
Chr.2: 13282355 - 13285678 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001084517.1 NP_001077986.1 3 - 4 619 92
NM_001336323.1 NP_001324768.1 1 - 2 540 92
NM_128673.5 NP_565717.2 3 - 4 627 92

View Full Product Details