Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HDG3
Entrez Gene ID
817798
Gene Name
homeodomain GLABROUS 3
Gene Aliases
AT2G32370, T32F6.11, T32F6_11, homeodomain GLABROUS 3
Chromosome Location
Chr.2: 13740878 - 13746286 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_128796.3 NP_180796.2 4 - 5 610 89
GenBank mRNA EF988635.1 - 4 - 5 610 89

View Full Product Details