Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RLP28
Entrez Gene ID
817870
Gene Name
receptor like protein 28
Gene Aliases
AT2G33080, AtRLP28, F25I18.18, F25I18_18, receptor like protein 28
Chromosome Location
Chr.2: 14031998 - 14034433 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_128868.2 NP_180867.1 1 - 1 2192 63

View Full Product Details