Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
iqd9
Entrez Gene ID
817960
Gene Name
IQ-domain 9
Gene Aliases
AT2G33990, IQ-domain 9, T14G11.11, T14G11_11
Chromosome Location
Chr.2: 14360255 - 14362511 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_128950.5 NP_180946.2 4 - 5 882 100
GenBank mRNA AK229389.1 - 4 - 5 549 100

View Full Product Details