Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
B ALPHA
Entrez Gene ID
841594
Gene Name
protein phosphatase 2A 55 kDa regulatory subunit B alpha
Gene Aliases
AT1G51690, ATB ALPHA, F19C24.10, F19C24_10, protein phosphatase 2A 55 kDa regulatory subunit B alpha isoform
Chromosome Location
Chr.1: 19163843 - 19170449 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001198270.1 NP_001185199.1 11 - 12 1418 124
NM_001333494.1 NP_001322512.1 9 - 10 3317 124
NM_001333495.1 NP_001322511.1 11 - 12 1439 124
NM_104047.4 NP_564595.1 10 - 11 3046 124
NM_202274.2 NP_974003.1 9 - 10 1340 124
GenBank mRNA AK230310.1 - 10 - 11 2826 124
AY120756.1 - 9 - 10 1341 124
BT000109.1 - 9 - 10 1099 124

View Full Product Details