Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PANB2
Entrez Gene ID
825326
Gene Name
Phosphoenolpyruvate carboxylase family protein
Gene Aliases
AT3G61530, KETOPANTOATE HYDROXYMETHYLTRANSFERASE, KPHMT2
Chromosome Location
Chr.3: 22771405 - 22773790 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_116018.4 NP_191712.1 2 - 3 664 109
NM_202745.4 NP_974474.1 3 - 4 1006 109
GenBank mRNA AY087528.1 - 2 - 3 664 109
BT025255.1 - 2 - 3 621 109

View Full Product Details