Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SDG14
Entrez Gene ID
825347
Gene Name
SET domain protein 14
Gene Aliases
AT3G61740, ATX3, F21F14.13, SET domain protein 14
Chromosome Location
Chr.3: 22850811 - 22857016 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001084857.2 NP_001078326.1 19 - 20 3058 104
NM_116039.4 NP_191733.3 18 - 19 3122 104
GenBank mRNA AB493660.1 - 18 - 19 2698 104

View Full Product Details