Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ATERDJ3B
Entrez Gene ID
825434
Gene Name
DNAJ heat shock family protein
Gene Aliases
AT3G62600, ERDJ3B
Chromosome Location
Chr.3: 23150779 - 23153649 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_116125.3 NP_191819.1 9 - 10 1187 115
GenBank mRNA AY087640.1 - 9 - 10 1048 115
AY094429.1 - 9 - 10 1017 115
AY125534.1 - 9 - 10 884 115

View Full Product Details