Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- AT-HSFA7B
- Entrez Gene ID
-
825510
- Gene Name
- winged-helix DNA-binding transcription factor family protein
- Gene Aliases
-
AT3G63350, HEAT SHOCK TRANSCRIPTION FACTOR A7B, HEAT SHOCK TRANSCRIPTION FACTOR A7B, HSFA7B, MAA21.4
- Chromosome Location
-
Chr.3: 23399236 - 23400812 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_116200.2 |
NP_191894.1 |
1 - 2 |
496 |
98 |
GenBank mRNA |
DQ446784.1 |
- |
1 - 2 |
264 |
98 |
View Full Product Details