Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- IND
- Entrez Gene ID
-
827911
- Gene Name
- basic helix-loop-helix (bHLH) DNA-binding superfamily protein
- Gene Aliases
-
AT4G00120, EDA33, EMBRYO SAC DEVELOPMENT ARREST 33, F6N15.18, F6N15_18, GT140, IND1, INDEHISCENT
- Chromosome Location
-
Chr.4: 41665 - 43197 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_116229.2 |
NP_191923.1 |
1 - 1 |
1335 |
87 |
View Full Product Details