Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ATSTE24
Entrez Gene ID
828209
Gene Name
Peptidase family M48 family protein
Gene Aliases
AT4G01320, F2N1.21, F2N1_21, STE24
Chromosome Location
Chr.4: 545796 - 549413 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001340307.1 NP_001329598.1 6 - 7 904 83
NM_001340308.1 NP_001329597.1 7 - 8 805 83
NM_116362.3 NP_567212.1 6 - 7 639 83
GenBank mRNA AY090232.1 - 6 - 7 606 83
BT006355.1 - 6 - 7 530 83

View Full Product Details