Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SWN
Entrez Gene ID
828165
Gene Name
SET domain-containing protein
Gene Aliases
AT4G02020, EZA1, SDG10, SET DOMAIN-CONTAINING PROTEIN 10, SWINGER, T10M13.3, T10M13_3
Chromosome Location
Chr.4: 886580 - 891987 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001340355.1 NP_001328437.1 4 - 5 682 70
NM_001340356.1 NP_001328436.1 4 - 5 679 70
NM_001340357.1 NP_001328435.1 4 - 5 679 70
NM_116433.3 NP_567221.1 4 - 5 699 70
GenBank mRNA AF100163.1 - 4 - 5 653 70
AY057477.1 - 4 - 5 679 70
AY090293.1 - 4 - 5 586 70

View Full Product Details