The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001203731.2 | NP_001190660.1 | 5 - 6 | 779 | 97 |
NM_116490.4 | NP_192165.1 | 4 - 5 | 922 | 97 | |
GenBank mRNA | AK226746.1 | - | 4 - 5 | 832 | 97 |