Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LD
Entrez Gene ID
827904
Gene Name
Homeodomain-like superfamily protein
Gene Aliases
AT4G02560, T10P11.15, T10P11_15, luminidependens
Chromosome Location
Chr.4: 1123475 - 1128490 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001203731.2 NP_001190660.1 10 - 11 1180 92
NM_116490.4 NP_192165.1 9 - 10 1323 92
GenBank mRNA AK226746.1 - 9 - 10 1233 92

View Full Product Details