Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
XTH9
Entrez Gene ID
828024
Gene Name
xyloglucan endotransglucosylase/hydrolase 9
Gene Aliases
AT4G03210, F4C21.14, F4C21_14, xyloglucan endotransglucosylase/hydrolase 9
Chromosome Location
Chr.4: 1415617 - 1417444 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001036504.1 NP_001031581.1 3 - 4 537 90
NM_116559.3 NP_192230.1 3 - 4 869 90
GenBank mRNA AB493675.1 - 3 - 4 467 90
AY044333.1 - 3 - 4 467 90
AY072353.1 - 3 - 4 534 90
BT002199.1 - 3 - 4 467 90

View Full Product Details