Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PETC
Entrez Gene ID
827996
Gene Name
photosynthetic electron transfer C
Gene Aliases
AT4G03280, F4C21.21, F4C21_21, PGR1, PROTON GRADIENT REGULATION 1, RIESKE IRON-SULFUR PROTEIN PRECURSOR, photosynthetic electron transfer C
Chromosome Location
Chr.4: 1440146 - 1441863 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_116566.4 NP_192237.1 4 - 5 760 119
NM_178964.2 NP_849295.1 3 - 4 820 119
GenBank mRNA AF370566.1 - 4 - 5 592 119
AF410296.1 - 3 - 4 823 119
AJ243702.1 - 4 - 5 697 119
AY093726.1 - 3 - 4 535 119

View Full Product Details