Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TPC1
Entrez Gene ID
825655
Gene Name
two-pore channel 1
Gene Aliases
AT4G03560, ATCCH1, ATTPC1, CALCIUM CHANNEL 1, FATTY ACID OXYGENATION UPREGULATED 2, FOU2, T5L23.5, two-pore channel 1, two-pore channel 1
Chromosome Location
Chr.4: 1579929 - 1585980 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_116594.5 NP_567258.1 18 - 19 2146 107
GenBank mRNA AF360372.1 - 18 - 19 1822 107
AK316725.1 - 18 - 19 1863 107

View Full Product Details