Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
WRKY42
Entrez Gene ID
825775
Gene Name
WRKY family transcription factor
Gene Aliases
AT4G04450, AtWRKY42, T26N6.6, T26N6_6
Chromosome Location
Chr.4: 2218042 - 2221260 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001340482.1 NP_001328851.1 1 - 2 969 76
NM_116683.3 NP_192354.1 2 - 3 671 76
GenBank mRNA AB493676.1 - 2 - 3 334 76
AY052650.1 - 2 - 3 334 76
DQ446799.1 - 2 - 3 334 76

View Full Product Details