Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CPK23
Entrez Gene ID
825809
Gene Name
calcium-dependent protein kinase 23
Gene Aliases
AT4G04740, ATCPK23, T4B21.15, T4B21_15, calcium-dependent protein kinase 23
Chromosome Location
Chr.4: 2402897 - 2408737 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001203743.2 NP_001190672.1 1 - 2 914 90
NM_001340501.1 NP_001319868.1 1 - 2 914 90
NM_001340502.1 NP_001328084.1 1 - 2 914 90
NM_001340503.1 NP_001328085.1 1 - 2 914 90
NM_001340504.1 NP_001328083.1 1 - 2 914 90
NM_001340505.1 NP_001328081.1 1 - 2 914 90
NM_001340506.1 NP_001328082.1 1 - 2 914 90
GenBank mRNA AY059823.1 - 1 - 2 742 90
AY081557.1 - 1 - 2 670 90

View Full Product Details