Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CPK23
Entrez Gene ID
825809
Gene Name
calcium-dependent protein kinase 23
Gene Aliases
AT4G04740, ATCPK23, T4B21.15, T4B21_15, calcium-dependent protein kinase 23
Chromosome Location
Chr.4: 2402897 - 2408737 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001203743.2 NP_001190672.1 9 - 10 2094 125
NM_001340501.1 NP_001319868.1 8 - 9 2578 125
NM_001340502.1 NP_001328084.1 8 - 9 2578 125
NM_001340505.1 NP_001328081.1 8 - 9 2578 125
GenBank mRNA AY059823.1 - 8 - 9 2406 125

View Full Product Details