Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SULTR3;5
Entrez Gene ID
832080
Gene Name
sulfate transporter 3;5
Gene Aliases
AT5G19600, T29J13.20, T29J13_20, sulfate transporter 3;5
Chromosome Location
Chr.5: 6612815 - 6617047 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_121965.4 NP_568377.1 8 - 9 1831 68
GenBank mRNA AB061739.1 - 8 - 9 1781 68
AK226785.1 - 8 - 9 1789 68

View Full Product Details