Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
OEP80
Entrez Gene ID
832082
Gene Name
outer envelope protein of 80 kDa
Gene Aliases
ARABIDOPSIS THALIANA OUTER ENVELOPE PROTEIN OF 80 KDA, AT5G19620, ATOEP80, EMB213, EMBRYO DEFECTIVE 213, T29J13.40, T29J13_40, TOC75, TOC75-V, outer envelope protein of 80 kDa, translocon at the outer envelope membrane of chloroplasts 75-V
Chromosome Location
Chr.5: 6623197 - 6627894 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_121967.4 NP_568378.1 15 - 16 2167 98
GenBank mRNA AF360205.1 - 15 - 16 2160 98
AY040053.1 - 15 - 16 2041 98

View Full Product Details