Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ATG12A
Entrez Gene ID
841861
Gene Name
Ubiquitin-like superfamily protein
Gene Aliases
APG12, AT1G54210, ATATG12, AUTOPHAGY 12, AUTOPHAGY 12 A, F20D21.38, F20D21_38
Chromosome Location
Chr.1: 20241226 - 20242641 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001198290.1 NP_001185219.1 3 - 4 246 137
NM_104299.3 NP_175823.1 5 - 6 373 137
GenBank mRNA AB073184.1 - 5 - 6 359 137
AF492758.1 - 5 - 6 330 137
AK176553.1 - 3 - 4 222 137

View Full Product Details