Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- PLDEPSILON
- Entrez Gene ID
-
841961
- Gene Name
- phospholipase D alpha 4
- Gene Aliases
-
AT1G55180, F7A10.25, F7A10_25, PLDALPHA4, phospholipase D alpha 4
- Chromosome Location
-
Chr.1: 20585008 - 20587723 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_104391.2 |
NP_175914.1 |
1 - 2 |
180 |
127 |
View Full Product Details