Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- HUB2
- Entrez Gene ID
-
841968
- Gene Name
- histone mono-ubiquitination 2
- Gene Aliases
-
AT1G55250, F7A10.17, F7A10_17, histone mono-ubiquitination 2
- Chromosome Location
-
Chr.1: 20606953 - 20612649 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_104398.4 |
NP_564680.4 |
3 - 4 |
428 |
97 |
View Full Product Details