Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CSLE1
Entrez Gene ID
842035
Gene Name
cellulose synthase like E1
Gene Aliases
AT1G55850, ATCSLE1, F14J16.9, F14J16_9, cellulose synthase like E1
Chromosome Location
Chr.1: 20876440 - 20879482 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_104462.4 NP_175981.2 4 - 5 1607 69
GenBank mRNA AK227116.1 - 4 - 5 1349 69
AY064040.1 - 4 - 5 1348 69
KJ138961.1 - 4 - 5 1295 69

View Full Product Details