Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SK42
Entrez Gene ID
842162
Gene Name
shaggy-like kinase 42
Gene Aliases
AT1G57870, ATSK42, F12K22.12, F12K22_12, shaggy-like kinase 42
Chromosome Location
Chr.1: 21431051 - 21436329 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001198327.1 NP_001185256.1 3 - 4 590 67
NM_001333808.1 NP_001319262.1 2 - 3 477 67
NM_001333809.1 NP_001320412.1 3 - 4 883 67
NM_104580.5 NP_176096.1 3 - 4 820 67
GenBank mRNA BT002018.1 - 3 - 4 593 67
BT008861.1 - 1 - 2 142 67

View Full Product Details