Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CW7
Entrez Gene ID
842242
Gene Name
CW7
Gene Aliases
AT1G59520, T30E16.4, T30E16_4
Chromosome Location
Chr.1: 21861282 - 21866210 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001333852.1 NP_001319271.1 10 - 11 1738 122
NM_001333854.1 NP_001321060.1 11 - 12 1342 122
NM_104645.4 NP_564743.1 11 - 12 1447 122
NM_179497.1 NP_849828.1 11 - 12 1345 122
GenBank mRNA AB028226.1 - 11 - 12 1252 122
AK227804.1 - 11 - 12 1329 122

View Full Product Details