Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CW7
Entrez Gene ID
842242
Gene Name
CW7
Gene Aliases
AT1G59520, T30E16.4, T30E16_4
Chromosome Location
Chr.1: 21861282 - 21866210 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001333852.1 NP_001319271.1 1 - 2 337 70
NM_001333853.1 NP_001321061.1 1 - 2 337 70
NM_001333854.1 NP_001321060.1 1 - 2 337 70
NM_104645.4 NP_564743.1 1 - 2 337 70
NM_179496.2 NP_849827.1 1 - 2 322 70
NM_179497.1 NP_849828.1 1 - 2 235 70
GenBank mRNA AB028226.1 - 1 - 2 142 70
AK227804.1 - 1 - 2 219 70
BT004825.1 - 1 - 2 30 70

View Full Product Details