Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CW7
Entrez Gene ID
842242
Gene Name
CW7
Gene Aliases
AT1G59520, T30E16.4, T30E16_4
Chromosome Location
Chr.1: 21861282 - 21866210 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001333852.1 NP_001319271.1 9 - 10 1097 69
NM_001333853.1 NP_001321061.1 9 - 10 1097 69
NM_001333854.1 NP_001321060.1 9 - 10 1097 69
NM_104645.4 NP_564743.1 9 - 10 1097 69
NM_179496.2 NP_849827.1 9 - 10 1082 69
NM_179497.1 NP_849828.1 9 - 10 995 69
GenBank mRNA AB028226.1 - 9 - 10 902 69
AK227804.1 - 9 - 10 979 69
BT004825.1 - 9 - 10 790 69

View Full Product Details