Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CW14
Entrez Gene ID
842255
Gene Name
CW14 protein (DUF1336)
Gene Aliases
AT1G59650, T30E16.22, T30E16_22
Chromosome Location
Chr.1: 21919745 - 21923013 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_104658.4 NP_564750.1 3 - 4 972 112
GenBank mRNA AB028233.1 - 3 - 4 972 112
BT004250.1 - 3 - 4 698 112
BT005503.1 - 3 - 4 618 112

View Full Product Details