Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ARF1
Entrez Gene ID
842268
Gene Name
auxin response factor 1
Gene Aliases
AT1G59750, F23H11.7, F23H11_7, auxin response factor 1
Chromosome Location
Chr.1: 21979289 - 21984433 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001036131.3 NP_001031208.1 5 - 6 423 102
NM_001198337.1 NP_001185266.1 5 - 6 669 102
NM_104668.4 NP_176184.1 5 - 6 619 102
NM_179499.2 NP_849830.1 5 - 6 574 102
GenBank mRNA AK317214.1 - 5 - 6 379 102
BT002748.1 - 4 - 5 308 102
U83245.1 - 5 - 6 422 102

View Full Product Details