Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
E1 ALPHA
Entrez Gene ID
842284
Gene Name
pyruvate dehydrogenase complex E1 alpha subunit
Gene Aliases
AT-E1 ALPHA, AT1G59900, PYRUVATE DEHYDROGENASE E1 ALPHA SUBUNIT, pyruvate dehydrogenase complex E1 alpha subunit
Chromosome Location
Chr.1: 22051133 - 22053885 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_104682.3 NP_176198.1 6 - 7 1355 94
GenBank mRNA AY087667.1 - 6 - 7 1180 94
BT000974.1 - 6 - 7 1120 94

View Full Product Details