Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- VPS34
- Entrez Gene ID
-
842344
- Gene Name
- vacuolar protein sorting 34
- Gene Aliases
-
AT1G60490, ATVPS34, F8A5.4, F8A5_4, PHOSPATIDYLINOSITOL 3-KINASE, PHOSPHATIDYLINOSITOL 3-KINASE, PI3K, vacuolar protein sorting 34
- Chromosome Location
-
Chr.1: 22285572 - 22290389 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_104735.4 |
NP_176251.1 |
3 - 4 |
508 |
63 |
GenBank mRNA |
BT000878.1 |
- |
3 - 4 |
309 |
63 |
View Full Product Details