Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CB5LP
Entrez Gene ID
842360
Gene Name
cytochrome B5-like protein
Gene Aliases
ARABIDOPSIS CYTOCHROME B5-LIKE PROTEIN, AT1G60660, ATCB5LP, B5 #5, F8A5.18, F8A5_18, cytochrome B5-like protein
Chromosome Location
Chr.1: 22342290 - 22343269 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_104749.3 NP_176265.1 1 - 1 594 62
GenBank mRNA AY049297.1 - 1 - 1 339 62
AY133575.1 - 1 - 1 279 62

View Full Product Details