Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AGL55
Entrez Gene ID
842383
Gene Name
AGAMOUS-like 55
Gene Aliases
AGAMOUS-like 55, AT1G60920, T7P1.6, T7P1_6
Chromosome Location
Chr.1: 22429692 - 22430361 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_104772.2 NP_176288.1 1 - 1 616 101

View Full Product Details