Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SCPL32
Entrez Gene ID
842406
Gene Name
serine carboxypeptidase-like 32
Gene Aliases
AT1G61130, F11P17.14, F11P17_14, serine carboxypeptidase-like 32
Chromosome Location
Chr.1: 22528853 - 22531827 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_104794.3 NP_176308.2 6 - 7 950 84
GenBank mRNA DQ056504.1 - 6 - 7 907 84

View Full Product Details