Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TPS14
Entrez Gene ID
842465
Gene Name
terpene synthase 14
Gene Aliases
AT1G61680, ATTPS14, T13M11.3, T13M11_3, TERPENE SYNTHASE 14, terpene synthase 14
Chromosome Location
Chr.1: 22772315 - 22774720 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001198357.1 NP_001185286.1 1 - 2 234 133
NM_104850.2 NP_176361.2 1 - 2 201 133

View Full Product Details