Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CPK19
Entrez Gene ID
842490
Gene Name
calcium-dependent protein kinase 19
Gene Aliases
AT1G61950, F8K4.14, F8K4_14, calcium-dependent protein kinase 19
Chromosome Location
Chr.1: 22899389 - 22902118 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_104875.3 NP_176386.2 2 - 3 786 86
GenBank mRNA DQ446385.1 - 2 - 3 758 86

View Full Product Details