Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- ALE1
- Entrez Gene ID
-
842532
- Gene Name
- PA-domain containing subtilase family protein
- Gene Aliases
-
ABNORMAL LEAF-SHAPE, ABNORMAL LEAF-SHAPE 1, ALE, AT1G62340, F24O1.36, F24O1_36
- Chromosome Location
-
Chr.1: 23051002 - 23055672 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_104914.2 |
NP_564793.2 |
8 - 9 |
1833 |
59 |
View Full Product Details