Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
WRKY19
Entrez Gene ID
826810
Gene Name
protein kinase family protein
Gene Aliases
AT4G12020, ATWRKY19, F16J13.90, F16J13_90, MAPK/ERK KINASE KINASE 4, MAPKKK11, MEKK4, MITOGEN-ACTIVATED PROTEIN KINASE KINASE KINASE 11
Chromosome Location
Chr.4: 7201522 - 7209692 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001125496.2 NP_001118968.1 4 - 5 1818 117
NM_001160750.1 NP_001154222.1 4 - 5 1818 117
NM_001340760.1 NP_001329658.1 4 - 5 1850 117
NM_001340761.1 NP_001329659.1 4 - 5 1818 117
NM_001340762.1 NP_001329652.1 4 - 5 1952 117
NM_001340763.1 NP_001329653.1 3 - 4 2071 117
NM_001340764.1 NP_001329654.1 4 - 5 1904 117
NM_001340765.1 NP_001329655.1 4 - 5 1904 117
NM_001340766.1 NP_001329656.1 4 - 5 1904 117
NM_001340767.1 NP_001329657.1 4 - 5 1893 117
NM_117272.3 NP_192939.2 4 - 5 1818 117

View Full Product Details