Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
WRKY19
Entrez Gene ID
826810
Gene Name
protein kinase family protein
Gene Aliases
AT4G12020, ATWRKY19, F16J13.90, F16J13_90, MAPK/ERK KINASE KINASE 4, MAPKKK11, MEKK4, MITOGEN-ACTIVATED PROTEIN KINASE KINASE KINASE 11
Chromosome Location
Chr.4: 7201522 - 7209692 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001125496.2 NP_001118968.1 8 - 9 3766 106
NM_001160750.1 NP_001154222.1 8 - 9 3766 106
NM_001340767.1 NP_001329657.1 8 - 9 3841 106
NM_117272.3 NP_192939.2 8 - 9 3766 106

View Full Product Details