Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PTAC12
Entrez Gene ID
818029
Gene Name
plastid transcriptionally active 12
Gene Aliases
AT2G34640, HEMERA, HMR, T31E10.2, T31E10_2, TAC12, plastid transcriptionally active 12
Chromosome Location
Chr.2: 14581835 - 14584797 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_129018.4 NP_181011.2 7 - 8 1604 74

View Full Product Details